siRNA Details
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 8 | 1944 | ||
NM_014955.2 | NP_055770.1 | 8 | 2087 | |||
NM_015935.4 | NP_057019.3 | 8 | 2412 | |||
XM_017001457.1 | XP_016856946.1 | 6 | 1687 | |||
XM_017001458.1 | XP_016856947.1 | 7 | 2364 | |||
XM_017001459.1 | XP_016856948.1 | 7 | 1914 | |||
GenBank mRNA | AB020666.2 | - | 8 | 2167 | ||
AB242174.1 | - | 8 | 2337 | |||
AB242175.1 | - | 8 | 2325 | |||
AK001172.1 | - | 6 | 1099 | |||
AK027552.1 | - | 8 | 2331 | |||
AK027621.1 | - | 8 | 2026 | |||
AK027886.1 | - | 7 | 1883 | |||
AK074552.1 | - | 4 | 1165 | |||
AK094818.1 | - | 1 | 1452 | |||
AK124482.1 | - | 1 | 2133 | |||
AK129561.1 | - | 7 | 1760 | |||
AK223493.1 | - | 8 | 2373 | |||
AK291740.1 | - | 8 | 2359 | |||
AK304471.1 | - | 9 | 2193 | |||
AL049669.1 | - | 8 | 1869 | |||
BC006814.1 | - | 5 | 962 | |||
BC029083.1 | - | 8 | 2166 | 4509086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
