siRNA Details
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 8 | 2922 | ||
NM_014955.2 | NP_055770.1 | 8 | 3065 | |||
NM_015935.4 | NP_057019.3 | 8 | 3390 | |||
XM_017001457.1 | XP_016856946.1 | 6 | 2665 | |||
XM_017001458.1 | XP_016856947.1 | 7 | 3342 | |||
XM_017001459.1 | XP_016856948.1 | 7 | 2892 | |||
GenBank mRNA | AB242175.1 | - | 8 | 3303 | ||
AK027621.1 | - | 8 | 3004 | |||
AK094818.1 | - | 1 | 2429 | |||
AK129561.1 | - | 7 | 2738 | |||
AL049669.1 | - | 8 | 2847 | |||
BX094025.1 | - | 1 | 710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
