siRNA Details
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014955.2 | NP_055770.1 | 2 | 653 | ||
NM_015935.4 | NP_057019.3 | 2 | 978 | |||
XM_017001455.1 | XP_016856944.1 | 1 | 472 | |||
XM_017001456.1 | XP_016856945.1 | 1 | 472 | |||
XM_017001457.1 | XP_016856946.1 | 1 | 472 | |||
XM_017001458.1 | XP_016856947.1 | 2 | 1095 | |||
XM_017001459.1 | XP_016856948.1 | 2 | 645 | |||
GenBank mRNA | AB020666.2 | - | 2 | 733 | ||
AB242174.1 | - | 2 | 903 | |||
AB242175.1 | - | 2 | 891 | |||
AF132936.1 | - | 2 | 903 | |||
AK027552.1 | - | 2 | 897 | |||
AK027621.1 | - | 2 | 592 | |||
AK027886.1 | - | 1 | 449 | |||
AK074552.1 | - | 1 | 382 | |||
AK129561.1 | - | 1 | 327 | |||
AK223493.1 | - | 2 | 939 | |||
AK291740.1 | - | 2 | 925 | |||
AK304471.1 | - | 3 | 759 | |||
BC029083.1 | - | 2 | 732 | 4509086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
