siRNA Details
Gene Symbol: SLC9B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300754.1 | NP_001287683.1 | 4 | 633 | ||
NM_001300756.1 | NP_001287685.1 | 4 | 999 | |||
NM_178833.5 | NP_849155.2 | 5 | 1170 | |||
XM_006714085.2 | XP_006714148.1 | 5 | 1169 | |||
XM_006714086.3 | XP_006714149.1 | 4 | 998 | |||
GenBank mRNA | AK172823.1 | - | 5 | 1170 | ||
AK297010.1 | - | 4 | 544 | |||
AK298506.1 | - | 3 | 316 | |||
AK299888.1 | - | 4 | 558 | |||
BC009732.1 | - | 2 | 238 | |||
BC047447.1 | - | 5 | 700 | 5295722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
