siRNA Details
Gene Symbol: SNX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021249.3 | NP_067072.3 | 13 | 1807 | ||
NM_152233.2 | NP_689419.2 | 14 | 1702 | |||
XM_006720224.1 | XP_006720287.1 | 13 | 1589 | |||
GenBank mRNA | AF109364.1 | - | 9 | 1185 | ||
AF121856.1 | - | 14 | 1693 | |||
AK021456.1 | - | 9 | 1189 | |||
AK124315.1 | - | 1 | 998 | |||
AK292250.1 | - | 14 | 1685 | |||
AK296213.1 | - | 15 | 1830 | |||
BC001798.1 | - | 13 | 1807 | 3354467 | ||
U83194.1 | - | 14 | 1670 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
