siRNA Details
Gene Symbol: SNX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021249.3 | NP_067072.3 | 11 | 1094 | ||
NM_152233.2 | NP_689419.2 | 12 | 989 | |||
XM_006720224.1 | XP_006720287.1 | 11 | 876 | |||
GenBank mRNA | AF109364.1 | - | 7 | 472 | ||
AF121856.1 | - | 12 | 980 | |||
AK021456.1 | - | 7 | 476 | |||
AK292250.1 | - | 12 | 972 | |||
AK296213.1 | - | 13 | 1117 | |||
AK308464.1 | - | 13 | 1117 | |||
BC001798.1 | - | 11 | 1094 | 3354467 | ||
BI870378.1 | - | 6 | 370 | |||
U83194.1 | - | 12 | 957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
