siRNA Details
Gene Symbol: CROCCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026752.1 | 5 | 688 | |||
GenBank mRNA | AK090414.1 | - | 10 | 1469 | ||
BC006312.2 | - | 5 | 735 | |||
BC010508.1 | - | 2 | 230 | |||
BC047736.1 | - | 5 | 688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
