siRNA Details
Gene Symbol: ZNF652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145365.1 | NP_001138837.1 | 2 | 1278 | ||
NM_014897.2 | NP_055712.1 | 2 | 1133 | |||
GenBank mRNA | AB023141.1 | - | 2 | 1133 | ||
AK292667.1 | - | 2 | 1268 | |||
BC139779.1 | - | 1 | 988 | 40122232 | ||
BC152440.1 | - | 2 | 1133 | 8862563 | ||
CR933680.1 | - | 2 | 1129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
