siRNA Details
Gene Symbol: ZNF652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145365.1 | NP_001138837.1 | 6 | 3137 | ||
NM_014897.2 | NP_055712.1 | 6 | 2992 | |||
XM_005257166.2 | XP_005257223.1 | 5 | 2344 | |||
GenBank mRNA | AB023141.1 | - | 6 | 2992 | ||
BC152440.1 | - | 6 | 2992 | 8862563 | ||
CR933680.1 | - | 6 | 2988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
