siRNA Details
Gene Symbol: FAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146094.1 | NP_001139566.1 | 2 | 1230 | ||
NM_001146095.1 | NP_001139567.1 | 2 | 1127 | |||
NM_001146096.1 | NP_001139568.1 | 2 | 1243 | |||
NM_014967.4 | NP_055782.3 | 2 | 1230 | |||
XM_005254232.4 | XP_005254289.1 | 2 | 1238 | |||
XM_005254234.4 | XP_005254291.1 | 2 | 1284 | |||
XM_005254235.3 | XP_005254292.1 | 2 | 1120 | |||
XM_011521371.1 | XP_011519673.1 | 2 | 1215 | |||
XM_011521372.2 | XP_011519674.1 | 2 | 1234 | |||
XM_017022012.1 | XP_016877501.1 | 2 | 1202 | |||
XM_017022013.1 | XP_016877502.1 | 2 | 1215 | |||
GenBank mRNA | AB023235.2 | - | 2 | 1177 | ||
AK290987.1 | - | 2 | 1210 | |||
BC047882.1 | - | 2 | 1130 | 5209981 | ||
BX647391.1 | - | 2 | 1224 | |||
CR936727.1 | - | 2 | 1210 | |||
CR936734.1 | - | 2 | 1200 | |||
HM560957.1 | - | 1 | 939 | |||
KJ534972.1 | - | 1 | 939 | |||
KJ534973.1 | - | 1 | 939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
