siRNA Details
Gene Symbol: FAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146094.1 | NP_001139566.1 | 3 | 1615 | ||
NM_001146095.1 | NP_001139567.1 | 3 | 1512 | |||
NM_001146096.1 | NP_001139568.1 | 3 | 1628 | |||
NM_014967.4 | NP_055782.3 | 3 | 1615 | |||
XM_005254232.4 | XP_005254289.1 | 3 | 1623 | |||
XM_005254234.4 | XP_005254291.1 | 3 | 1669 | |||
XM_005254235.3 | XP_005254292.1 | 3 | 1505 | |||
XM_011521370.2 | XP_011519672.1 | 3 | 308 | |||
XM_011521371.1 | XP_011519673.1 | 4 | 1734 | |||
XM_011521372.2 | XP_011519674.1 | 3 | 1619 | |||
XM_017022012.1 | XP_016877501.1 | 3 | 1587 | |||
XM_017022013.1 | XP_016877502.1 | 3 | 1600 | |||
GenBank mRNA | AB023235.2 | - | 3 | 1562 | ||
AK290987.1 | - | 3 | 1595 | |||
BC047882.1 | - | 3 | 1515 | 5209981 | ||
BX647391.1 | - | 3 | 1609 | |||
CR936727.1 | - | 4 | 1729 | |||
CR936734.1 | - | 3 | 1585 | |||
HM560957.1 | - | 2 | 1324 | |||
KJ534972.1 | - | 2 | 1324 | |||
KJ534973.1 | - | 2 | 1324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
