siRNA Details
Gene Symbol: FAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014967.4 | NP_055782.3 | 5 | 2056 | ||
XM_005254232.4 | XP_005254289.1 | 5 | 2064 | |||
XM_005254234.4 | XP_005254291.1 | 5 | 2110 | |||
XM_005254235.3 | XP_005254292.1 | 5 | 1946 | |||
XM_011521370.2 | XP_011519672.1 | 5 | 749 | |||
XM_011521371.1 | XP_011519673.1 | 6 | 2175 | |||
XM_011521372.2 | XP_011519674.1 | 5 | 2060 | |||
XM_017022012.1 | XP_016877501.1 | 4 | 1826 | |||
XM_017022013.1 | XP_016877502.1 | 4 | 1839 | |||
GenBank mRNA | AB023235.2 | - | 5 | 2003 | ||
CR936727.1 | - | 6 | 2170 | |||
CR936734.1 | - | 5 | 2026 | |||
HM560957.1 | - | 4 | 1765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
