siRNA Details
Gene Symbol: CD247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000734.3 | NP_000725.1 | 8 | 1488 | ||
NM_198053.2 | NP_932170.1 | 8 | 1491 | |||
XM_011510144.2 | XP_011508446.1 | 9 | 1559 | |||
XM_011510145.1 | XP_011508447.1 | 9 | 1565 | |||
XM_017002800.1 | XP_016858289.1 | 9 | 1550 | |||
XM_017002801.1 | XP_016858290.1 | 9 | 1545 | |||
GenBank mRNA | AK223325.1 | - | 8 | 1480 | ||
EF364118.1 | - | 1 | 166 | |||
J04132.1 | - | 8 | 1417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
