siRNA Details
Gene Symbol: Zbtb20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285805.1 | NP_001272734.1 | 11 | 2789 | ||
NM_019778.2 | NP_062752.2 | 9 | 2736 | |||
NM_181058.1 | NP_851401.1 | 7 | 2514 | |||
GenBank mRNA | AB221632.1 | - | 3 | 2201 | ||
AF185576.1 | - | 7 | 2559 | |||
AF194030.1 | - | 5 | 2351 | |||
AK005028.2 | - | 7 | 2515 | |||
AY028963.1 | - | 4 | 2318 | |||
BC031114.1 | - | 8 | 2622 | 4971586 | ||
BC056446.1 | - | 11 | 2761 | 5703319 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001105880.
Gene Ontology Categories:
