siRNA Details
Gene Symbol: ANKRD36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310154.1 | NP_001297083.1 | 43, 44 | 3333 | ||
GenBank mRNA | AK123626.1 | - | 7, 8 | 1039 | ||
AL832836.1 | - | 21, 22 | 1056 | |||
AL832977.1 | - | 14, 15 | 733 | |||
U28831.1 | - | 1, 2 | 64 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
