siRNA Details
Gene Symbol: ATP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320408.1 | NP_001307337.1 | 1 | 134 | ||
NM_001320409.1 | NP_001307338.1 | 1 | 134 | |||
NM_001320410.1 | NP_001307339.1 | 1 | 134 | |||
NM_033276.3 | NP_150592.1 | 1 | 134 | |||
XM_017020206.1 | XP_016875695.1 | 1 | 33 | |||
GenBank mRNA | AF078164.2 | - | 1 | 64 | ||
BC033881.1 | - | 1 | 74 | |||
BM544500.1 | - | 1 | 93 | |||
BU601924.1 | - | 1 | 137 | |||
DA711361.1 | - | 1 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
