siRNA Details
Gene Symbol: SLC22A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091990.1 | - | 8 | 958 | ||
AY517500.1 | - | 8 | 958 | |||
BC101315.1 | - | 7 | 1147 | |||
BC101317.1 | - | 4 | 724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
