siRNA Details
Gene Symbol: HMGN2P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022014.1 | 6 | 745 | |||
GenBank mRNA | AK096745.1 | - | 6 | 741 | ||
AY271964.1 | - | 6 | 731 | |||
AY271965.1 | - | 4 | 500 | |||
AY271966.1 | - | 4 | 506 | |||
AY271967.1 | - | 4 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
