siRNA Details
Gene Symbol: CYB5R4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016230.3 | NP_057314.2 | 2, 3 | 369 | ||
GenBank mRNA | AF169803.1 | - | 1, 2 | 141 | ||
AK313146.1 | - | 2, 3 | 342 | |||
AL832113.1 | - | 2, 3 | 334 | |||
BC025380.1 | - | 2, 3 | 336 | 4829042 | ||
DA274558.1 | - | 2, 3 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
