siRNA Details
Gene Symbol: ZNF322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242797.1 | NP_001229726.1 | 5 | 2062 | ||
NM_001242798.1 | NP_001229727.1 | 3 | 1917 | |||
NM_001242799.1 | NP_001229728.1 | 3 | 1898 | |||
NM_024639.4 | NP_078915.2 | 4 | 1987 | |||
GenBank mRNA | AK027046.1 | - | 1 | 1307 | ||
AY376736.1 | - | 4 | 1770 | |||
BC050425.1 | - | 1 | 1483 | 6473688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
