siRNA Details
Gene Symbol: ACTL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024675.1 | NP_001019846.1 | 1 | 707 | ||
GenBank mRNA | BC137111.1 | - | 1 | 687 | 9021108 | |
BC137114.1 | - | 1 | 687 | 9021111 | ||
BI832107.1 | - | 1 | 718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
