siRNA Details
Gene Symbol: TMEM89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008269.1 | NP_001008270.1 | 1 | 17 | ||
GenBank mRNA | BC133034.1 | - | 1 | 116 | 40147056 | |
BC133036.1 | - | 1 | 116 | 40147058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
