siRNA Details
Gene Symbol: PRKAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164758.1 | NP_001158230.1 | 2, 3 | 297 | ||
NM_001164759.1 | NP_001158231.1 | 2, 3 | 232 | |||
NM_001164760.1 | NP_001158232.1 | 2, 3 | 342 | |||
NM_001164761.1 | NP_001158233.1 | 2, 3 | 363 | |||
NM_001164762.1 | NP_001158234.1 | 2, 3 | 303 | |||
NM_002735.2 | NP_002726.1 | 2, 3 | 321 | |||
XM_011515446.2 | XP_011513748.1 | 1, 2 | 279 | |||
GenBank mRNA | AK098822.1 | - | 1, 2 | 263 | ||
AK297337.1 | - | 2, 3 | 273 | |||
AK315951.1 | - | 2, 3 | 333 | |||
AK315990.1 | - | 2, 3 | 293 | |||
AL833563.1 | - | 2, 3 | 263 | |||
BC026734.1 | - | 2, 3 | 271 | 5141110 | ||
BC036828.1 | - | 2, 3 | 294 | 5247772 | ||
CT003793.1 | - | 2, 3 | 342 | |||
DA162030.1 | - | 2, 3 | 363 | |||
DA210679.1 | - | 2, 3 | 233 | |||
DA501689.1 | - | 2, 3 | 304 | |||
DA910871.1 | - | 2, 3 | 297 | |||
DC314672.1 | - | 2, 3 | 321 | |||
M65066.1 | - | 1, 2 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
