siRNA Details
Gene Symbol: KRTAP4-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322457.1 | NP_001309386.1 | 1 | 538 | ||
NM_031960.2 | NP_114166.1 | 1 | 507 | |||
GenBank mRNA | AJ406940.1 | - | 1 | 255 |
Gene Symbol: KRTAP4-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033059.3 | NP_149048.2 | 1 | 537 |
Gene Symbol: KRTAP4-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031854.2 | NP_114060.1 | 1 | 561 | ||
GenBank mRNA | AJ406943.1 | - | 1 | 561 | ||
BC004180.2 | - | 1 | 541 | 2959821 | ||
BC004212.2 | - | 1 | 541 | 2959821 | ||
BC101159.1 | - | 1 | 553 | 40015747 | ||
BC101160.1 | - | 1 | 553 | 40015748 | ||
BC101162.1 | - | 1 | 553 | 40015750 |
Gene Symbol: KRTAP4-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033188.3 | NP_149445.3 | 1 | 491 | ||
GenBank mRNA | AJ406937.1 | - | 1 | 506 |
Gene Symbol: KRTAP4-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030976.1 | NP_112238.1 | 1 | 528 | ||
GenBank mRNA | AJ406938.1 | - | 1 | 227 | ||
AJ406945.1 | - | 1 | 492 |
Gene Symbol: KRTAP4-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033061.3 | NP_149050.3 | 1 | 378 |
Gene Symbol: KRTAP4-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146041.1 | NP_001139513.1 | 1 | 543 | ||
GenBank mRNA | AJ406941.1 | - | 1 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
