siRNA Details
Gene Symbol: TMEM229A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136002.1 | NP_001129474.1 | 1 | 1250 | ||
GenBank mRNA | BC157828.1 | - | 1 | 801 | 9057047 | |
BC171809.1 | - | 1 | 801 | 9054463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
