siRNA Details
Gene Symbol: CLDN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002026.2 | NP_001002026.1 | 5 | 941 | ||
NM_016369.3 | NP_057453.1 | 5 | 950 | |||
GenBank mRNA | AK098474.1 | - | 5 | 941 | ||
AK298103.1 | - | 6 | 973 | |||
AY358479.1 | - | 5 | 977 | |||
BC142708.1 | - | 5 | 956 | 40148818 | ||
BC146668.1 | - | 5 | 956 | 40148169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
