siRNA Details
Gene Symbol: CLDN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002026.2 | NP_001002026.1 | 5 | 1492 | ||
NM_016369.3 | NP_057453.1 | 5 | 1501 | |||
GenBank mRNA | AK098474.1 | - | 5 | 1492 | ||
AY358479.1 | - | 5 | 1528 | |||
BC142708.1 | - | 5 | 1507 | 40148818 | ||
BC146668.1 | - | 5 | 1507 | 40148169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
