siRNA Details
Gene Symbol: CLDN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002026.2 | NP_001002026.1 | 3 | 495 | ||
NM_016369.3 | NP_057453.1 | 3 | 504 | |||
GenBank mRNA | AF221069.1 | - | 3 | 451 | ||
AF349452.1 | - | 3 | 451 | |||
AK098474.1 | - | 3 | 495 | |||
AK298103.1 | - | 4 | 527 | |||
AY358479.1 | - | 3 | 531 | |||
BC142708.1 | - | 3 | 510 | 40148818 | ||
BC146668.1 | - | 3 | 510 | 40148169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
