siRNA Details
Gene Symbol: MARCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005415.1 | NP_001005415.1 | 3 | 479 | ||
NM_001005416.1 | NP_001005416.1 | 3 | 479 | |||
NM_016496.4 | NP_057580.3 | 4 | 750 | |||
XM_006722763.3 | XP_006722826.1 | 3 | 422 | |||
XM_011528046.2 | XP_011526348.1 | 3 | 424 | |||
XM_011528047.2 | XP_011526349.1 | 3 | 510 | |||
XM_017026853.1 | XP_016882342.1 | 4 | 975 | |||
GenBank mRNA | AB197929.1 | - | 2 | 360 | ||
AF151074.1 | - | 4 | 660 | |||
AK130163.1 | - | 3 | 468 | |||
BC015910.1 | - | 2 | 290 | |||
BC032624.1 | - | 3 | 764 | 5241282 | ||
BC111388.1 | - | 3 | 459 | 5242152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
