siRNA Details
Gene Symbol: MARCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005415.1 | NP_001005415.1 | 5 | 1311 | ||
NM_001005416.1 | NP_001005416.1 | 4 | 1101 | |||
NM_016496.4 | NP_057580.3 | 6 | 1582 | |||
XM_006722763.3 | XP_006722826.1 | 4 | 1044 | |||
XM_011528046.2 | XP_011526348.1 | 5 | 1286 | |||
XM_011528047.2 | XP_011526349.1 | 5 | 1372 | |||
XM_017026853.1 | XP_016882342.1 | 6 | 1807 | |||
XM_017026854.1 | XP_016882343.1 | 3 | 859 | |||
GenBank mRNA | AB197929.1 | - | 4 | 1191 | ||
AF151074.1 | - | 6 | 1492 | |||
AK130163.1 | - | 5 | 1300 | |||
BC015910.1 | - | 4 | 1123 | |||
BC032624.1 | - | 5 | 1597 | 5241282 | ||
BC111388.1 | - | 5 | 1290 | 5242152 | ||
CN363445.1 | - | 2 | 600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
