siRNA Details
Gene Symbol: TRIOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039141.2 | NP_001034230.1 | 14, 15 | 5856 | ||
NM_007032.5 | NP_008963.3 | 4, 5 | 450 | |||
NM_138632.2 | NP_619538.2 | 4, 5 | 450 | |||
GenBank mRNA | AF281030.1 | - | 4, 5 | 434 | ||
BC003618.2 | - | 4, 5 | 372 | |||
BC022200.1 | - | 4, 5 | 444 | |||
BQ880866.1 | - | 450 | ||||
BX342673.2 | - | 4, 5 | 444 | |||
DQ228003.1 | - | 12, 13 | 5519 | |||
DQ228005.1 | - | 13, 14 | 5721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
