siRNA Details
Gene Symbol: TRIOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039141.2 | NP_001034230.1 | 16 | 6347 | ||
NM_007032.5 | NP_008963.3 | 6 | 941 | |||
NM_138632.2 | NP_619538.2 | 6 | 941 | |||
GenBank mRNA | AF281030.1 | - | 6 | 925 | ||
AL713698.1 | - | 1 | 43 | |||
BC003618.2 | - | 6 | 863 | |||
BC004303.1 | - | 1 | 331 | |||
BC013278.2 | - | 1 | 118 | |||
BC022200.1 | - | 6 | 935 | |||
DQ228003.1 | - | 14 | 6010 | |||
DQ228005.1 | - | 15 | 6212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
