siRNA Details
Gene Symbol: TLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016170.4 | NP_057254.1 | 3 | 1617 | ||
GenBank mRNA | AB593086.1 | - | 4 | 1304 | ||
BC006356.2 | - | 3 | 1602 | 4125397 | ||
BC006396.1 | - | 2 | 1874 | |||
BC018611.1 | - | 4 | 1213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
