siRNA Details
Gene Symbol: GREB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014668.3 | NP_055483.2 | 33 | 6773 | ||
XM_005246192.4 | XP_005246249.1 | 33 | 6795 | |||
XM_005246196.3 | XP_005246253.1 | 17 | 3848 | |||
XM_011510418.2 | XP_011508720.1 | 33 | 6631 | |||
XM_011510419.2 | XP_011508721.1 | 32 | 6631 | |||
XM_011510422.2 | XP_011508724.1 | 18 | 3943 | |||
GenBank mRNA | AB011147.1 | - | 16 | 3576 | ||
BC054502.1 | - | 16 | 3585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
