siRNA Details
Gene Symbol: GRHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195010.1 | NP_001181939.1 | 4 | 560 | ||
NM_021180.3 | NP_067003.2 | 4 | 456 | |||
NM_198173.2 | NP_937816.1 | 4 | 625 | |||
NM_198174.2 | NP_937817.3 | 4 | 625 | |||
XM_011541869.1 | XP_011540171.1 | 4 | 506 | |||
XM_011541870.2 | XP_011540172.1 | 3 | 427 | |||
GenBank mRNA | AK074386.1 | - | 4 | 522 | ||
AK315164.1 | - | 4 | 427 | |||
AY231160.1 | - | 4 | 456 | |||
AY231161.1 | - | 3 | 269 | |||
BC036890.2 | - | 4 | 571 | 5502632 | ||
BU848928.1 | - | 4 | 556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
