siRNA Details
Gene Symbol: STAG3L5P-PVRIG2P-PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036569.1 | 14 | 2410 | |||
NR_036570.1 | 13 | 1943 | ||||
GenBank mRNA | AF217981.1 | - | 5 | 1348 | ||
AJ400846.1 | - | 3 | 250 | |||
AK056412.1 | - | 14 | 2361 | |||
AK074039.1 | - | 5 | 3162 | |||
AL834317.1 | - | 13 | 1765 | |||
AL834336.1 | - | 1 | 79 | |||
DQ851871.1 | - | 9 | 999 | |||
DQ851876.1 | - | 7 | 1503 | |||
DQ851877.1 | - | 7 | 1503 | |||
DQ851878.1 | - | 8 | 1092 | |||
DQ851880.1 | - | 6 | 863 | |||
DQ851881.1 | - | 5 | 802 |
Gene Symbol: PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC050547.1 | - | 3 | 212 | 5755993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
