siRNA Details
Gene Symbol: MATN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317748.1 | NP_001304677.1 | 10 | 1777 | ||
NM_002380.4 | NP_002371.3 | 11 | 1900 | |||
NM_030583.3 | NP_085072.2 | 11 | 1900 | |||
XM_005250920.1 | XP_005250977.1 | 11 | 1865 | |||
XM_017013417.1 | XP_016868906.1 | 10 | 1742 | |||
XM_017013418.1 | XP_016868907.1 | 11 | 1865 | |||
GenBank mRNA | AK027775.1 | - | 5 | 531 | ||
AK289721.1 | - | 11 | 2081 | |||
AL137638.1 | - | 9 | 962 | |||
AL833931.1 | - | 11 | 1788 | |||
AY358895.1 | - | 10 | 1687 | |||
BC010444.1 | - | 11 | 1829 | 4215380 | ||
BC016394.1 | - | 8 | 885 | |||
BX649093.1 | - | 8 | 954 | |||
U69263.2 | - | 10 | 1732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
