siRNA Details
Gene Symbol: MATN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317748.1 | NP_001304677.1 | 3 | 797 | ||
NM_002380.4 | NP_002371.3 | 3 | 797 | |||
NM_030583.3 | NP_085072.2 | 3 | 797 | |||
XM_005250920.1 | XP_005250977.1 | 3 | 762 | |||
XM_017013417.1 | XP_016868906.1 | 3 | 762 | |||
XM_017013418.1 | XP_016868907.1 | 3 | 762 | |||
GenBank mRNA | AK075489.1 | - | 3 | 705 | ||
AL833931.1 | - | 3 | 685 | |||
AY358895.1 | - | 3 | 707 | |||
BC010444.1 | - | 3 | 726 | 4215380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
