siRNA Details
Gene Symbol: SOGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080627.3 | NP_542194.2 | 14 | 3858 | ||
NM_199181.2 | NP_954650.2 | 14 | 3131 | |||
GenBank mRNA | AB020696.1 | - | 1 | 312 | ||
AK126630.1 | - | 14 | 3131 | |||
AK131491.1 | - | 10 | 2358 | |||
BC113405.1 | - | 1 | 262 | |||
BC113433.1 | - | 1 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
