siRNA Details
Gene Symbol: XPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022459.4 | NP_071904.4 | 23 | 4930 | ||
XM_005266502.1 | XP_005266559.1 | 24 | 5085 | |||
XM_005266503.3 | XP_005266560.1 | 20 | 4588 | |||
XM_005266504.3 | XP_005266561.1 | 17 | 4457 | |||
XM_011535197.1 | XP_011533499.1 | 24 | 5094 | |||
XM_011535198.1 | XP_011533500.1 | 22 | 4772 | |||
XM_011535199.1 | XP_011533501.1 | 21 | 4735 | |||
XM_011535200.2 | XP_011533502.1 | 21 | 6185 | |||
XM_017020704.1 | XP_016876193.1 | 19 | 4726 | |||
XM_017020705.1 | XP_016876194.1 | 20 | 5962 | |||
XM_017020706.1 | XP_016876195.1 | 17 | 4081 | |||
GenBank mRNA | AB051508.1 | - | 23 | 4893 | ||
AL831819.2 | - | 6 | 2234 | |||
BX647843.1 | - | 14 | 3702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
