siRNA Details
Gene Symbol: KIAA0513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286565.1 | NP_001273494.1 | 3 | 1041 | ||
NM_001286566.1 | NP_001273495.1 | 3 | 690 | |||
NM_001297766.1 | NP_001284695.1 | 3 | 683 | |||
NM_014732.3 | NP_055547.1 | 3 | 683 | |||
XM_005256265.3 | XP_005256322.1 | 3 | 732 | |||
XM_017023912.1 | XP_016879401.1 | 3 | 3258 | |||
GenBank mRNA | AB011085.1 | - | 3 | 1041 | ||
AK299892.1 | - | 3 | 674 | |||
BC030280.1 | - | 3 | 630 | 5211581 | ||
BU844795.1 | - | 3 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
