siRNA Details
Gene Symbol: MYO1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033054.2 | NP_149043.2 | 19 | 2697 | ||
XM_017012503.1 | XP_016867992.1 | 18 | 2456 | |||
XM_017012504.1 | XP_016867993.1 | 19 | 2692 | |||
GenBank mRNA | AB290179.1 | - | 19 | 2657 | ||
AK074135.1 | - | 14 | 2861 | |||
AK131089.1 | - | 14 | 4814 | |||
AL832302.1 | - | 18 | 2452 | |||
BC015693.2 | - | 3 | 387 | |||
BC113544.1 | - | 19 | 2644 | 8322596 | ||
BC117241.1 | - | 19 | 2644 | 40125792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
