siRNA Details
Gene Symbol: MMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100600.1 | NP_001094070.1 | 7 | 1647 | ||
NM_001270375.1 | NP_001257304.1 | 8 | 1625 | |||
NM_198403.3 | NP_940685.3 | 7 | 1575 | |||
GenBank mRNA | BC037881.1 | - | 7 | 1598 | 5274201 | |
BC067905.1 | - | 7 | 1575 | 5312325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
