siRNA Details
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 6 | 756 | ||
NM_001265611.1 | NP_001252540.1 | 6 | 756 | |||
NM_014994.2 | NP_055809.2 | 6 | 756 | |||
NR_049761.1 | 6 | 756 | ||||
NR_049762.1 | 5 | 664 | ||||
XM_006720438.1 | XP_006720501.1 | 6 | 720 | |||
XM_011521383.1 | XP_011519685.1 | 6 | 720 | |||
XM_011521384.2 | XP_011519686.1 | 6 | 720 | |||
XM_017022017.1 | XP_016877506.1 | 5 | 996 | |||
GenBank mRNA | AB011168.2 | - | 6 | 666 | ||
AK091616.1 | - | 6 | 642 | |||
AK292414.1 | - | 6 | 627 | |||
AK302482.1 | - | 5 | 605 | |||
AL833267.1 | - | 6 | 676 | |||
BC036660.1 | - | 6 | 761 | 5266613 | ||
BC113983.1 | - | 6 | 648 | 40083186 | ||
BC114493.1 | - | 6 | 648 | 40083182 | ||
BX640787.1 | - | 5 | 438 | |||
BX648786.1 | - | 6 | 707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
