siRNA Details
Gene Symbol: CLUAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015041.2 | NP_055856.1 | 2 | 217 | ||
XM_017023068.1 | XP_016878557.1 | 2 | 232 | |||
XM_017023069.1 | XP_016878558.1 | 2 | 232 | |||
XM_017023070.1 | XP_016878559.1 | 2 | 217 | |||
XM_017023071.1 | XP_016878560.1 | 2 | 1206 | |||
XM_017023072.1 | XP_016878561.1 | 2 | 1206 | |||
GenBank mRNA | AB014543.1 | - | 2 | 144 | ||
AB089691.1 | - | 2 | 199 | |||
BC017070.2 | - | 2 | 145 | 3916900 | ||
DA502615.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
