siRNA Details
Gene Symbol: CLUAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015041.2 | NP_055856.1 | 7 | 817 | ||
NM_024793.2 | NP_079069.1 | 3 | 275 | |||
XM_006720867.3 | XP_006720930.1 | 6 | 668 | |||
XM_017023068.1 | XP_016878557.1 | 7 | 832 | |||
XM_017023069.1 | XP_016878558.1 | 7 | 832 | |||
XM_017023070.1 | XP_016878559.1 | 7 | 817 | |||
XM_017023071.1 | XP_016878560.1 | 7 | 1806 | |||
XM_017023072.1 | XP_016878561.1 | 7 | 1806 | |||
XM_017023073.1 | XP_016878562.1 | 6 | 668 | |||
GenBank mRNA | AB014543.1 | - | 7 | 744 | ||
AB089691.1 | - | 7 | 799 | |||
AK023319.1 | - | 1 | 33 | |||
AK023359.1 | - | 3 | 275 | |||
AK294318.1 | - | 1 | 89 | |||
BC017070.2 | - | 7 | 745 | 3916900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
