siRNA Details
Gene Symbol: GSDMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166237.1 | NP_001159709.1 | 6 | 1055 | ||
NM_024736.6 | NP_079012.3 | 3 | 501 | |||
XM_011517301.1 | XP_011515603.1 | 3 | 470 | |||
GenBank mRNA | AF318341.1 | - | 4 | 539 | ||
AK022212.1 | - | 3 | 460 | |||
AK123985.1 | - | 2 | 684 | |||
AK127941.1 | - | 2 | 898 | |||
AK291817.1 | - | 6 | 1055 | |||
AK303615.1 | - | 3 | 462 | |||
AY008304.1 | - | 3 | 490 | |||
BC008904.2 | - | 3 | 486 | 3634992 | ||
BC069000.1 | - | 3 | 441 | 6213121 | ||
DC398191.1 | - | 3 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
