siRNA Details
Gene Symbol: GSDMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166237.1 | NP_001159709.1 | 5 | 873 | ||
NM_024736.6 | NP_079012.3 | 2 | 319 | |||
XM_011517301.1 | XP_011515603.1 | 2 | 288 | |||
GenBank mRNA | AF318341.1 | - | 3 | 357 | ||
AK022212.1 | - | 2 | 278 | |||
AK123985.1 | - | 2 | 278 | |||
AK127941.1 | - | 1 | 716 | |||
AK291817.1 | - | 5 | 873 | |||
AK303615.1 | - | 2 | 280 | |||
AY008304.1 | - | 2 | 308 | |||
BC008904.2 | - | 2 | 304 | 3634992 | ||
BC069000.1 | - | 2 | 259 | 6213121 | ||
DC398191.1 | - | 2 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
