siRNA Details
Gene Symbol: CHD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170629.1 | NP_001164100.1 | 13 | 2959 | ||
NM_020920.3 | NP_065971.2 | 14 | 2204 | |||
GenBank mRNA | AB046784.3 | - | 13 | 2447 | ||
AK122868.1 | - | 4 | 569 | |||
AK122910.1 | - | 14 | 2151 | |||
CR749315.1 | - | 14 | 2149 |

More Information
Other Species Targeted

Gene Ontology Categories:
