siRNA Details
Gene Symbol: CHD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170629.1 | NP_001164100.1 | 19 | 4069 | ||
NM_020920.3 | NP_065971.2 | 20 | 3314 | |||
GenBank mRNA | AB046784.3 | - | 19 | 3557 | ||
AK098408.1 | - | 4 | 679 | |||
AK122868.1 | - | 10 | 1679 | |||
AK122910.1 | - | 20 | 3262 | |||
AK131077.1 | - | 5 | 893 | |||
CR749315.1 | - | 20 | 3259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
